siRNA Details
| Gene Symbol: CARD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032587.3 | NP_115976.2 | 2 | 686 | ||
| XM_017009990.1 | XP_016865479.1 | 2 | 804 | |||
| GenBank mRNA | AB209844.1 | - | 2 | 506 | ||
| AF356193.1 | - | 2 | 810 | |||
| AK304349.1 | - | 2 | 618 | |||
| AY196783.1 | - | 2 | 686 | |||
| BC093825.1 | - | 2 | 633 | 7939670 | ||

CARD6
Genomic Map