siRNA Details
| Gene Symbol: GPR146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303473.1 | NP_001290402.1 | 2 | 1775 | ||
| NM_001303474.1 | NP_001290403.1 | 3 | 2043 | |||
| NM_138445.3 | NP_612454.1 | 2 | 1898 | |||
| XM_005249613.2 | XP_005249670.1 | 3 | 2452 | |||
| XM_006715642.2 | XP_006715705.1 | 3 | 2620 | |||
| XM_006715643.2 | XP_006715706.1 | 3 | 2570 | |||
| XM_011515116.2 | XP_011513418.1 | 3 | 2017 | |||
| GenBank mRNA | AK300166.1 | - | 2 | 1481 | ||
| BC014241.2 | - | 1 | 1676 | 4563636 | ||
More Information
Set Membership: |
|

GPR146
Genomic Map