siRNA Details
Gene Symbol: ADRA2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000683.3 | NP_000674.2 | 1 | 1721 | ||
GenBank mRNA | BC142625.1 | - | 1 | 1892 | 40147492 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
