siRNA Details
| Gene Symbol: PSD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012455.2 | NP_036587.2 | 2 | 734 | ||
| XM_005263634.2 | XP_005263691.1 | 2 | 799 | |||
| XM_006712392.2 | XP_006712455.1 | 2 | 746 | |||
| XM_006712393.2 | XP_006712456.1 | 2 | 746 | |||
| XM_006712394.2 | XP_006712457.1 | 2 | 746 | |||
| GenBank mRNA | AB208798.1 | - | 2 | 737 | ||
| AJ459781.1 | - | 1 | 572 | |||
| AK290045.1 | - | 2 | 744 | |||
| BC035307.1 | - | 2 | 720 | 4339380 | ||
| BC073151.1 | - | 2 | 696 | 5757431 | ||
| BQ061445.1 | - | 2 | 735 | |||
| U63127.1 | - | 1 | 614 | |||

PSD4
Genomic Map