siRNA Details
| Gene Symbol: CYP26C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183374.2 | NP_899230.2 | 4 | 770 | ||
| GenBank mRNA | AK127234.1 | - | 1 | 1616 | ||
| AY356349.1 | - | 4 | 770 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYP26C1| Gene Symbol: CYP26C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183374.2 | NP_899230.2 | 4 | 770 | ||
| GenBank mRNA | AK127234.1 | - | 1 | 1616 | ||
| AY356349.1 | - | 4 | 770 | |||