siRNA Details
| Gene Symbol: OR4D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004707.3 | NP_001004707.1 | 1 | 769 | ||
| GenBank mRNA | AF303373.1 | - | 1 | 1024 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR4D2| Gene Symbol: OR4D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004707.3 | NP_001004707.1 | 1 | 769 | ||
| GenBank mRNA | AF303373.1 | - | 1 | 1024 | ||