siRNA Details
| Gene Symbol: CLEC5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013252.2 | NP_037384.1 | 4, 5 | 399 | ||
| XM_017011915.1 | XP_016867404.1 | 3, 4 | 357 | |||
| XM_017011916.1 | XP_016867405.1 | 4, 5 | 425 | |||
| GenBank mRNA | AF139768.1 | - | 4, 5 | 353 | ||
| AJ271684.1 | - | 4, 5 | 399 | |||
| AK292766.1 | - | 4, 5 | 388 | |||
| AK313703.1 | - | 4, 5 | 388 | |||
| BC093714.1 | - | 3, 4 | 223 | 7939559 | ||
| BC112099.1 | - | 3, 4 | 223 | 8327567 | ||
| BC113098.1 | - | 4, 5 | 341 | 40076723 | ||

CLEC5A
Genomic Map