siRNA Details
| Gene Symbol: ZDHHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013304.2 | NP_037436.1 | 10 | 1389 | ||
| XM_011523060.1 | XP_011521362.1 | 8 | 922 | |||
| GenBank mRNA | BC021908.1 | - | 10 | 1415 | 4053374 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZDHHC1| Gene Symbol: ZDHHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013304.2 | NP_037436.1 | 10 | 1389 | ||
| XM_011523060.1 | XP_011521362.1 | 8 | 922 | |||
| GenBank mRNA | BC021908.1 | - | 10 | 1415 | 4053374 | |