siRNA Details
| Gene Symbol: WBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012478.3 | NP_036610.2 | 8 | 1023 | ||
| XM_011524580.2 | XP_011522882.1 | 7 | 809 | |||
| GenBank mRNA | AF087866.1 | - | 8 | 927 | ||
| AK057881.1 | - | 8 | 928 | |||
| AK094458.1 | - | 2 | 1149 | |||
| AK128407.1 | - | 4 | 2841 | |||
| AK293214.1 | - | 9 | 1077 | |||
| AK294082.1 | - | 7 | 803 | |||
| AK294155.1 | - | 6 | 633 | |||
| AL049981.1 | - | 2 | 203 | |||
| BC007452.1 | - | 8 | 926 | 3140484 | ||
| BC010616.1 | - | 8 | 936 | 4177319 | ||

WBP2
Genomic Map