siRNA Details
| Gene Symbol: MGAT4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC064141.1 | - | 3 | 1284 | 30378187 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGAT4C| Gene Symbol: MGAT4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC064141.1 | - | 3 | 1284 | 30378187 | |