siRNA Details
| Gene Symbol: ZNF180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278508.2 | NP_001265437.2 | 4 | 2942 | ||
| NM_001278509.2 | NP_001265438.2 | 5 | 2945 | |||
| NM_001288759.2 | NP_001275688.2 | 5 | 3022 | |||
| NM_001288760.2 | NP_001275689.1 | 7 | 3190 | |||
| NM_001288761.2 | NP_001275690.1 | 4 | 2818 | |||
| NM_001288762.2 | NP_001275691.1 | 4 | 2895 | |||
| NM_001291633.1 | NP_001278562.1 | 5 | 2940 | |||
| NM_013256.5 | NP_037388.3 | 5 | 3017 | |||
| XM_011527280.2 | XP_011525582.1 | 5 | 5350 | |||
| XM_017027250.1 | XP_016882739.1 | 5 | 3091 | |||
| XM_017027251.1 | XP_016882740.1 | 6 | 3160 | |||
| XM_017027252.1 | XP_016882741.1 | 6 | 3066 | |||
| XM_017027253.1 | XP_016882742.1 | 6 | 3135 | |||
| XM_017027254.1 | XP_016882743.1 | 4 | 2701 | |||
| XM_017027255.1 | XP_016882744.1 | 6 | 3153 | |||
| XM_017027256.1 | XP_016882745.1 | 6 | 3205 | |||
| GenBank mRNA | AF192913.1 | - | 6 | 2997 | ||
| AK024047.1 | - | 5 | 2943 | |||
| AK122688.1 | - | 4 | 2939 | |||

ZNF180
Genomic Map