siRNA Details
Gene Symbol: MYLPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324458.1 | NP_001311387.1 | 8, 9 | 625 | ||
NM_001324459.1 | NP_001311388.1 | 8, 9 | 653 | |||
NM_013292.4 | NP_037424.2 | 6, 7 | 488 | |||
GenBank mRNA | AF363061.1 | - | 6, 7 | 461 | ||
AK314082.1 | - | 6, 7 | 488 | |||
BC012571.1 | - | 6, 7 | 466 | 4245958 | ||
BX649054.1 | - | 6, 7 | 456 | |||
CR456963.1 | - | 5, 6 | 407 | |||
M21812.1 | - | 6, 7 | 468 | |||
X57543.1 | - | 6, 7 | 468 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
