siRNA Details
| Gene Symbol: ST8SIA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001307986.1 | NP_001294915.1 | 8 | 2800 | ||
| NM_001307987.1 | NP_001294916.1 | 6 | 2599 | |||
| NM_013305.5 | NP_037437.2 | 7 | 2692 | |||
| XM_017025716.1 | XP_016881205.1 | 7 | 2366 | |||

ST8SIA5
Genomic Map