siRNA Details
Gene Symbol: GPR132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278694.1 | NP_001265623.1 | 5 | 1478 | ||
NM_001278695.1 | NP_001265624.1 | 3 | 608 | |||
NM_001278696.1 | NP_001265625.1 | 2 | 494 | |||
NM_013345.3 | NP_037477.1 | 4 | 1388 | |||
GenBank mRNA | AB465599.1 | - | 4 | 1268 | ||
AB465600.1 | - | 3 | 488 | |||
AF083955.1 | - | 4 | 1256 | |||
AK292142.1 | - | 5 | 1351 | |||
AK303654.1 | - | 3 | 488 | |||
AX775841.1 | - | 3 | 503 | |||
AX775935.1 | - | 3 | 488 | |||
AX775939.1 | - | 3 | 966 | |||
BC084546.1 | - | 2 | 943 | 5455247 | ||
DB137292.1 | - | 2 | 374 | |||
EU432121.1 | - | 2 | 356 | |||
EU543705.1 | - | 4 | 477 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
