siRNA Details
| Gene Symbol: ZNF221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297588.1 | NP_001284517.1 | 4 | 440 | ||
| NM_001297589.1 | NP_001284518.1 | 4 | 450 | |||
| NM_013359.2 | NP_037491.2 | 5 | 588 | |||
| XM_017027232.1 | XP_016882721.1 | 4 | 467 | |||
| XM_017027233.1 | XP_016882722.1 | 3 | 384 | |||
| GenBank mRNA | AF187987.1 | - | 5 | 588 | ||
| AK314207.1 | - | 4 | 435 | |||
| BC111981.1 | - | 4 | 343 | 8327449 | ||
| BC113426.1 | - | 4 | 343 | 8322478 | ||
| BC143659.1 | - | 5 | 491 | |||
| DA493401.1 | - | 4 | 441 | |||

ZNF221
Genomic Map