siRNA Details
| Gene Symbol: ZNF222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129996.1 | NP_001123468.1 | 4 | 407 | ||
| NM_013360.2 | NP_037492.2 | 4 | 375 | |||
| XM_005259208.3 | XP_005259265.1 | 5 | 471 | |||
| XM_005259209.3 | XP_005259266.1 | 3 | 328 | |||
| GenBank mRNA | AF187988.1 | - | 4 | 354 | ||
| AK095196.1 | - | 6 | 567 | |||
| BC030261.1 | - | 4 | 349 | 5210539 | ||
| DB185422.1 | - | 5 | 536 | |||

ZNF222
Genomic Map