siRNA Details
| Gene Symbol: ZNF225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321685.1 | NP_001308614.1 | 4 | 538 | ||
| NM_013362.3 | NP_037494.2 | 3 | 373 | |||
| XM_011527285.2 | XP_011525587.1 | 3 | 393 | |||
| XM_011527286.2 | XP_011525588.1 | 3 | 1037 | |||
| GenBank mRNA | AF187991.1 | - | 3 | 347 | ||
| AK223477.1 | - | 3 | 292 | |||
| AK292437.1 | - | 3 | 377 | |||
| AK302106.1 | - | 2 | 89 | |||
| AK313301.1 | - | 2 | 380 | |||
| BC108912.2 | - | 2 | 170 | 40027157 | ||
| DA537943.1 | - | 3 | 340 | |||

ZNF225
Genomic Map