siRNA Details
| Gene Symbol: WDR37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014023.3 | NP_054742.2 | 2, 3 | 314 | ||
| GenBank mRNA | AB023199.1 | - | 2, 3 | 285 | ||
| AK292591.1 | - | 2, 3 | 277 | |||
| AL136827.1 | - | 2, 3 | 292 | |||
| BC018044.1 | - | 2, 3 | 343 | 4794184 | ||

WDR37
Genomic Map