siRNA Details
| Gene Symbol: GPR132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278694.1 | NP_001265623.1 | 5 | 1679 | ||
| NM_001278695.1 | NP_001265624.1 | 3 | 809 | |||
| NM_001278696.1 | NP_001265625.1 | 2 | 695 | |||
| NM_013345.3 | NP_037477.1 | 4 | 1589 | |||
| GenBank mRNA | AB465599.1 | - | 4 | 1469 | ||
| AB465600.1 | - | 3 | 689 | |||
| AF083955.1 | - | 4 | 1457 | |||
| AK292142.1 | - | 5 | 1552 | |||
| AK303654.1 | - | 3 | 689 | |||
| AX775841.1 | - | 3 | 704 | |||
| AX775935.1 | - | 3 | 689 | |||
| AX775939.1 | - | 3 | 1167 | |||
| BC084546.1 | - | 2 | 1144 | 5455247 | ||
| EU432121.1 | - | 2 | 557 | |||
| EU543705.1 | - | 4 | 678 | |||
More Information
Set Membership: |
|

GPR132
Genomic Map