siRNA Details
| Gene Symbol: RPA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013347.4 | NP_037479.1 | 1 | 756 | ||
| GenBank mRNA | BC069791.1 | - | 1 | 651 | 7262610 | |
| BC069808.1 | - | 1 | 651 | 7262634 | ||
| BC069824.1 | - | 1 | 651 | 7262622 | ||
| BC104013.2 | - | 1 | 627 | 40024430 | ||
| BC104014.2 | - | 1 | 627 | 40024431 | ||
| U24186.2 | - | 1 | 757 | |||

RPA4
Genomic Map