siRNA Details
| Gene Symbol: ZNF223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF187989.1 | - | 5 | 645 | ||
| AK292327.1 | - | 5 | 623 | |||
| BC022466.1 | - | 5 | 660 | 4793666 | ||
| Gene Symbol: ZNF284 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK308295.1 | - | 7 | 820 | ||

ZNF223
Genomic Map