siRNA Details
| Gene Symbol: TMEM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135820.1 | NP_001129292.1 | 3 | 956 | ||
| NM_013390.2 | NP_037522.1 | 3 | 956 | |||
| XM_005251869.4 | XP_005251926.1 | 3 | 589 | |||
| GenBank mRNA | AB037833.2 | - | 3 | 549 | ||
| AF137030.1 | - | 3 | 564 | |||
| AK300405.1 | - | 3 | 585 | |||
| BC140773.1 | - | 3 | 458 | 9021641 | ||
| BC144510.1 | - | 3 | 458 | |||
| BC146780.1 | - | 3 | 549 | 9007233 | ||
| BC171782.1 | - | 3 | 458 | |||

TMEM2
Genomic Map