siRNA Details
| Gene Symbol: SACM1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319072.1 | NP_001306001.1 | 5, 6 | 648 | ||
| NM_001319073.1 | NP_001306002.1 | 7, 8 | 816 | |||
| NM_014016.4 | NP_054735.3 | 6, 7 | 776 | |||
| XM_011533500.2 | XP_011531802.1 | 6, 7 | 567 | |||
| GenBank mRNA | AB020658.1 | - | 6, 7 | 620 | ||
| AK291142.1 | - | 6, 7 | 660 | |||
| AK296421.1 | - | 5, 6 | 430 | |||
| AK301248.1 | - | 7, 8 | 656 | |||
| AL136831.1 | - | 6, 7 | 566 | |||
| BC016559.1 | - | 6, 7 | 572 | 3049075 | ||
| CR749564.1 | - | 6, 7 | 1722 | |||

SACM1L
Genomic Map