siRNA Details
| Gene Symbol: WDR37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014023.3 | NP_054742.2 | 4 | 461 | ||
| GenBank mRNA | AB023199.1 | - | 4 | 432 | ||
| AK128194.1 | - | 3 | 3270 | |||
| AK292591.1 | - | 4 | 424 | |||
| AK307259.1 | - | 3 | 354 | |||
| AL136827.1 | - | 4 | 439 | |||
| BC018044.1 | - | 4 | 490 | 4794184 | ||

WDR37
Genomic Map