siRNA Details
| Gene Symbol: METTL7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014033.3 | NP_054752.3 | 2 | 2832 | ||
| XM_006719332.1 | XP_006719395.1 | 3 | 3186 | |||
| GenBank mRNA | AJ536233.1 | - | 3 | 1149 | ||
| AK023693.1 | - | 3 | 1159 | |||
| AK024409.1 | - | 3 | 1877 | |||
| AK130665.1 | - | 1 | 1514 | |||
| AK225595.1 | - | 3 | 1455 | |||
| AL050159.1 | - | 3 | 1368 | |||
| AY358797.1 | - | 3 | 1277 | |||

METTL7A
Genomic Map