siRNA Details
| Gene Symbol: HCK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172129.1 | NP_001165600.1 | 4 | 482 | ||
| NM_001172130.1 | NP_001165601.1 | 4 | 479 | |||
| NM_001172131.1 | NP_001165602.1 | 4 | 479 | |||
| NM_001172132.1 | NP_001165603.1 | 5 | 548 | |||
| NM_001172133.1 | NP_001165604.1 | 5 | 546 | |||
| NM_002110.3 | NP_002101.2 | 4 | 482 | |||
| GenBank mRNA | AK026432.1 | - | 4 | 416 | ||
| AK225819.1 | - | 4 | 428 | |||
| AK289896.1 | - | 4 | 470 | |||
| AK290928.1 | - | 5 | 494 | |||
| AK298726.1 | - | 5 | 492 | |||
| AK314335.1 | - | 4 | 395 | |||
| BC014435.1 | - | 4 | 368 | 4855747 | ||
| BC094847.1 | - | 4 | 379 | 5923744 | ||
| BC108930.1 | - | 4 | 409 | 40025497 | ||
| BC108931.1 | - | 4 | 409 | 40025500 | ||
| BC113854.1 | - | 4 | 406 | 40025503 | ||
| BC114463.1 | - | 4 | 409 | 40025502 | ||
| M16592.1 | - | 3 | 257 | |||

HCK
Genomic Map