siRNA Details
| Gene Symbol: ANAPC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286756.1 | NP_001273685.1 | 16 | 1334 | ||
| NM_013367.2 | NP_037499.2 | 16 | 1334 | |||
| XM_005248159.2 | XP_005248216.1 | 3 | 228 | |||
| XM_011513838.1 | XP_011512140.1 | 16 | 1211 | |||
| XM_011513839.1 | XP_011512141.1 | 16 | 1255 | |||
| XM_017008138.1 | XP_016863627.1 | 16 | 1263 | |||
| GenBank mRNA | AF191338.1 | - | 15 | 1205 | ||
| AK023597.1 | - | 13 | 945 | |||
| AK292336.1 | - | 16 | 1254 | |||
| BC059383.1 | - | 16 | 1307 | 30349524 | ||

ANAPC4
Genomic Map