siRNA Details
| Gene Symbol: DDAH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303007.1 | NP_001289936.1 | 6 | 2034 | ||
| NM_001303008.1 | NP_001289937.1 | 7 | 1134 | |||
| NM_013974.2 | NP_039268.1 | 7 | 1209 | |||
| XM_011514448.2 | XP_011512750.1 | 7 | 1285 | |||
| GenBank mRNA | AF070667.1 | - | 7 | 1206 | ||
| AF087894.1 | - | 7 | 1206 | |||
| AK026191.1 | - | 6 | 1561 | |||
| AK098671.1 | - | 7 | 1196 | |||
| BC001435.2 | - | 7 | 1121 | 3138832 | ||
| FJ224290.1 | - | 7 | 1206 | |||

DDAH2
Genomic Map