siRNA Details
| Gene Symbol: DEXI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014015.3 | NP_054734.2 | 2 | 1468 | ||
| XM_011522455.2 | XP_011520757.1 | 3 | 850 | |||
| XM_017023171.1 | XP_016878660.1 | 3 | 2236 | |||
| GenBank mRNA | AF108145.2 | - | 2 | 1084 | ||
| AF129929.1 | - | 2 | 980 | |||
| BC001083.2 | - | 2 | 1204 | 2958243 | ||
| BC037569.1 | - | 2 | 1468 | 5227606 | ||
| BC052989.1 | - | 2 | 1201 | 6194689 | ||
| BC071683.1 | - | 2 | 1367 | 5190179 | ||

DEXI
Genomic Map