siRNA Details
| Gene Symbol: DUOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014080.4 | NP_054799.4 | 8, 9 | 1154 | ||
| XM_005254421.2 | XP_005254478.1 | 8, 9 | 1154 | |||
| GenBank mRNA | AB209010.1 | - | 5, 6 | 1282 | ||
| AF230496.1 | - | 8, 9 | 1136 | |||
| AF267981.1 | - | 8, 9 | 1154 | |||

DUOX2
Genomic Map