siRNA Details
| Gene Symbol: HIGD1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099668.1 | NP_001093138.1 | 4 | 1456 | ||
| NM_001099669.1 | NP_001093139.1 | 4 | 1309 | |||
| NM_014056.3 | NP_054775.2 | 4 | 1285 | |||
| GenBank mRNA | AF077034.1 | - | 4 | 1238 | ||
| AF145385.1 | - | 4 | 1224 | |||
| AK225571.1 | - | 4 | 1240 | |||
| AL110233.1 | - | 4 | 1255 | |||
| BC000601.1 | - | 4 | 1223 | 3346355 | ||
| BC009583.1 | - | 4 | 1216 | 4093421 | ||
| BC009594.1 | - | 4 | 1222 | 4109190 | ||
| BC070277.1 | - | 4 | 1238 | 4686431 | ||

HIGD1A
Genomic Map