siRNA Details
| Gene Symbol: OGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014057.4 | NP_054776.1 | 7 | 1513 | ||
| NM_024416.4 | NP_077727.3 | 7 | 1432 | |||
| NM_033014.3 | NP_148935.1 | 7 | 1795 | |||
| GenBank mRNA | AF086912.1 | - | 7 | 1360 | ||
| AF100758.1 | - | 7 | 1361 | |||
| AK075521.1 | - | 6 | 1324 | |||
| AK129845.1 | - | 6 | 1317 | |||
| AL110267.1 | - | 6 | 1302 | |||
| AL133114.1 | - | 3 | 778 | |||
| BC037273.2 | - | 6 | 1301 | 5265880 | ||
| BC095443.1 | - | 6 | 1317 | 30405707 | ||

OGN
Genomic Map