siRNA Details
| Gene Symbol: WDR91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014149.3 | NP_054868.3 | 14 | 2101 | ||
| XM_011516083.2 | XP_011514385.1 | 15 | 2206 | |||
| XM_011516086.2 | XP_011514388.1 | 15 | 2128 | |||
| XM_017012070.1 | XP_016867559.1 | 14 | 2149 | |||
| XM_017012071.1 | XP_016867560.1 | 12 | 1641 | |||
| XM_017012072.1 | XP_016867561.1 | 14 | 2083 | |||
| GenBank mRNA | AF161534.1 | - | 14 | 2028 | ||
| AK001606.1 | - | 14 | 2056 | |||
| AK022935.1 | - | 13 | 1932 | |||
| AK074466.1 | - | 7 | 861 | |||
| AK124765.1 | - | 1 | 2180 | |||
| AL136940.1 | - | 14 | 1952 | |||
| BC009939.2 | - | 12 | 1559 | |||
| BC017246.2 | - | 14 | 2040 | 4890904 | ||
| BC063394.1 | - | 14 | 2037 | |||
| BX640957.1 | - | 14 | 3021 | |||
| CR533561.1 | - | 13 | 1883 | |||

WDR91
Genomic Map