siRNA Details
| Gene Symbol: CCDC59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014167.4 | NP_054886.2 | 4 | 657 | ||
| NR_033192.1 | 4 | 586 | ||||
| GenBank mRNA | AF161477.1 | - | 4 | 631 | ||
| AF213377.1 | - | 4 | 624 | |||
| AL832749.1 | - | 2 | 1677 | |||
| BC020647.1 | - | 4 | 620 | 4771954 | ||
| BM846580.1 | - | 4 | 556 | |||

CCDC59
Genomic Map