siRNA Details
| Gene Symbol: TM4SF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014220.2 | NP_055035.1 | 3 | 549 | ||
| XM_017006385.1 | XP_016861874.1 | 3 | 549 | |||
| GenBank mRNA | AK093829.1 | - | 1 | 172 | ||
| AK130073.1 | - | 3 | 461 | |||
| AK130271.1 | - | 3 | 543 | |||
| AL832780.1 | - | 1 | 644 | |||
| BC008442.1 | - | 3 | 416 | 4101110 | ||
| BC010166.2 | - | 3 | 365 | 4040623 | ||
| BC034145.1 | - | 3 | 510 | 4618019 | ||
| CR456953.1 | - | 3 | 315 | |||
| M90657.1 | - | 423 | ||||

TM4SF1
Genomic Map