siRNA Details
| Gene Symbol: SLC6A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014229.2 | NP_055044.1 | 5, 6 | 825 | ||
| XM_011534033.2 | XP_011532335.1 | 5, 6 | 825 | |||
| XM_017007073.1 | XP_016862562.1 | 6, 7 | 676 | |||
| GenBank mRNA | AK312719.1 | - | 5, 6 | 825 | ||
| S75989.1 | - | 5, 6 | 793 | |||

SLC6A11
Genomic Map