siRNA Details
| Gene Symbol: TMEM50A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014313.3 | NP_055128.1 | 3 | 283 | ||
| XM_005245817.1 | XP_005245874.1 | 3 | 247 | |||
| XM_011541159.1 | XP_011539461.1 | 3 | 477 | |||
| GenBank mRNA | AF081282.1 | - | 3 | 210 | ||
| AF087880.1 | - | 3 | 271 | |||
| AK299180.1 | - | 3 | 247 | |||
| AK312588.1 | - | 3 | 247 | |||
| AL136627.1 | - | 3 | 265 | |||
| AY071927.1 | - | 3 | 260 | |||
| AY358650.1 | - | 3 | 246 | |||
| BC007341.2 | - | 3 | 231 | 3688672 | ||
| BC066908.1 | - | 3 | 222 | 4817784 | ||
| JX644905.1 | - | 3 | 283 | |||
| JX644906.1 | - | 3 | 283 | |||

TMEM50A
Genomic Map