siRNA Details
| Gene Symbol: CARHSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042476.2 | NP_001035941.1 | 3 | 590 | ||
| NM_001278260.1 | NP_001265189.1 | 4 | 398 | |||
| NM_001278261.1 | NP_001265190.1 | 4 | 537 | |||
| NM_001278262.1 | NP_001265191.1 | 4 | 496 | |||
| NM_001278263.1 | NP_001265192.1 | 3 | 272 | |||
| NM_001278264.1 | NP_001265193.1 | 3 | 414 | |||
| NM_001278265.1 | NP_001265194.1 | 3 | 406 | |||
| NM_001278266.1 | NP_001265195.1 | 3 | 382 | |||
| NM_014316.3 | NP_055131.2 | 3 | 284 | |||
| GenBank mRNA | AF115345.1 | - | 2 | 241 | ||
| AK093698.1 | - | 2 | 976 | |||
| AK311777.1 | - | 3 | 278 | |||
| BC001333.1 | - | 3 | 590 | |||
| BC003366.1 | - | 3 | 281 | 2900952 | ||
| BC108283.1 | - | 3 | 264 | 4430943 | ||
| BM693675.1 | - | 3 | 414 | |||
| BM920489.1 | - | 3 | 272 | |||
| CA396216.1 | - | 3 | 341 | |||
| DA736976.1 | - | 4 | 538 | |||
| DB101255.1 | - | 4 | 392 | |||
| HY005510.1 | - | 3 | 407 | |||
| HY090855.1 | - | 3 | 284 | |||

CARHSP1
Genomic Map