siRNA Details
| Gene Symbol: RUSC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105203.1 | NP_001098673.1 | 9 | 2759 | ||
| NM_001105204.1 | NP_001098674.1 | 9 | 2441 | |||
| NM_001105205.1 | NP_001098675.1 | 8 | 1781 | |||
| NM_001278227.1 | NP_001265156.1 | 8 | 1725 | |||
| NM_001278228.1 | NP_001265157.1 | 7 | 1961 | |||
| NM_001278229.1 | NP_001265158.1 | 8 | 1506 | |||
| NM_001278230.1 | NP_001265159.1 | 7 | 1623 | |||
| NM_014328.4 | NP_055143.2 | 8 | 1824 | |||
| NR_103478.1 | 7 | 1171 | ||||
| XM_006711254.1 | XP_006711317.1 | 10 | 3170 | |||
| XM_006711256.1 | XP_006711319.1 | 10 | 2852 | |||
| XM_006711257.1 | XP_006711320.1 | 8 | 1438 | |||
| XM_017000891.1 | XP_016856380.1 | 9 | 3340 | |||
| XM_017000892.1 | XP_016856381.1 | 8 | 1120 | |||
| GenBank mRNA | AB026894.2 | - | 8 | 1248 | ||
| AK055451.1 | - | 8 | 2019 | |||
| AK074485.1 | - | 7 | 1059 | |||
| AK074982.1 | - | 8 | 1243 | |||
| AK093777.1 | - | 6 | 2083 | |||
| AK125378.1 | - | 9 | 2393 | |||
| AK298730.1 | - | 8 | 1235 | |||
| AK307736.1 | - | 7 | 1410 | |||
| AK314559.1 | - | 8 | 1493 | |||
| AL080083.1 | - | 7 | 1101 | |||
| BC001045.2 | - | 8 | 1265 | |||
| BC004293.1 | - | 7 | 1027 | |||
| BC028055.1 | - | 7 | 1373 | |||
| BC052277.1 | - | 8 | 1781 | |||
| BX402179.2 | - | 8 | 995 | |||
| BX640612.1 | - | 7 | 1441 | |||
| CR749671.1 | - | 7 | 2330 | |||

RUSC1
Genomic Map