siRNA Details
| Gene Symbol: MRPL22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014990.2 | NP_001014990.1 | 4 | 213 | ||
| NM_014180.3 | NP_054899.2 | 5 | 331 | |||
| GenBank mRNA | AF161507.1 | - | 5 | 299 | ||
| AK000601.1 | - | 5 | 318 | |||
| AK097880.1 | - | 1 | 1968 | |||
| AK302784.1 | - | 4 | 193 | |||
| BC012565.1 | - | 5 | 301 | 4333117 | ||
| BP372420.1 | - | 5 | 331 | |||
| BU943770.1 | - | 5 | 324 | |||
| CR933679.1 | - | 4 | 192 | |||

MRPL22
Genomic Map