siRNA Details
| Gene Symbol: EVI2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003927.2 | NP_001003927.1 | 3 | 628 | ||
| NM_014210.3 | NP_055025.2 | 2 | 487 | |||
| GenBank mRNA | AK295311.1 | - | 3 | 563 | ||
| AK312348.1 | - | 2 | 440 | |||
| AV724051.1 | - | 2 | 408 | |||
| BC035572.1 | - | 2 | 375 | 5229926 | ||
| BG699778.1 | - | 3 | 528 | |||
| BI597493.1 | - | 2 | 443 | |||
| DA364537.1 | - | 2 | 487 | |||

EVI2A
Genomic Map