siRNA Details
| Gene Symbol: HABP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014282.3 | NP_055097.2 | 2 | 579 | ||
| XM_005251812.3 | XP_005251869.1 | 2 | 410 | |||
| XM_017014484.1 | XP_016869973.1 | 2 | 241 | |||
| GenBank mRNA | AF241831.1 | - | 2 | 529 | ||
| BC018788.1 | - | 2 | 533 | 3502216 | ||
| BQ962055.1 | - | 2 | 294 | |||
| DR006985.1 | - | 2 | 580 | |||
| U77327.1 | - | 1 | 112 | |||

HABP4
Genomic Map