siRNA Details
| Gene Symbol: CAPN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014296.2 | NP_055111.1 | 1, 2 | 380 | ||
| XM_011533555.2 | XP_011531857.1 | 1, 2 | 231 | |||
| XM_011533556.2 | XP_011531858.1 | 2, 3 | 463 | |||
| XM_011533557.2 | XP_011531859.1 | 1, 2 | 353 | |||
| XM_017006043.1 | XP_016861532.1 | 2, 3 | 463 | |||
| XM_017006044.1 | XP_016861533.1 | 1, 2 | 353 | |||
| XM_017006045.1 | XP_016861534.1 | 2, 3 | 419 | |||
| GenBank mRNA | AB028639.1 | - | 1, 2 | 361 | ||
| AK314253.1 | - | 1, 2 | 342 | |||
| BC056202.1 | - | 1, 2 | 358 | 5274889 | ||
| BI464220.1 | - | 2, 3 | 577 | |||

CAPN7
Genomic Map