siRNA Details
| Gene Symbol: CARHSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042476.2 | NP_001035941.1 | 4 | 678 | ||
| NM_001278260.1 | NP_001265189.1 | 5 | 486 | |||
| NM_001278261.1 | NP_001265190.1 | 5 | 625 | |||
| NM_001278262.1 | NP_001265191.1 | 5 | 584 | |||
| NM_001278263.1 | NP_001265192.1 | 4 | 360 | |||
| NM_001278264.1 | NP_001265193.1 | 4 | 502 | |||
| NM_001278265.1 | NP_001265194.1 | 4 | 494 | |||
| NM_001278266.1 | NP_001265195.1 | 4 | 470 | |||
| NM_014316.3 | NP_055131.2 | 4 | 372 | |||
| XM_005255228.4 | XP_005255285.3 | 4 | 536 | |||
| XM_005255229.4 | XP_005255286.1 | 4 | 348 | |||
| XM_011522444.2 | XP_011520746.1 | 4 | 347 | |||
| GenBank mRNA | AF115345.1 | - | 3 | 329 | ||
| AK311777.1 | - | 4 | 366 | |||
| BC001333.1 | - | 4 | 678 | |||
| BC003366.1 | - | 4 | 369 | 2900952 | ||
| BC108283.1 | - | 4 | 352 | 4430943 | ||
| BM693675.1 | - | 4 | 502 | |||
| BM920489.1 | - | 4 | 360 | |||
| CA396216.1 | - | 4 | 429 | |||
| DB101255.1 | - | 5 | 480 | |||
| HY005510.1 | - | 4 | 495 | |||
| HY090855.1 | - | 4 | 372 | |||

CARHSP1
Genomic Map