siRNA Details
| Gene Symbol: MRPL22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014990.2 | NP_001014990.1 | 5 | 320 | ||
| NM_014180.3 | NP_054899.2 | 6 | 438 | |||
| GenBank mRNA | AF161507.1 | - | 6 | 406 | ||
| AK000601.1 | - | 6 | 425 | |||
| AK097880.1 | - | 2 | 2075 | |||
| AK302784.1 | - | 5 | 300 | |||
| BC012565.1 | - | 6 | 408 | 4333117 | ||
| BP372420.1 | - | 6 | 438 | |||
| BU943770.1 | - | 6 | 431 | |||
| CR933679.1 | - | 5 | 299 | |||

MRPL22
Genomic Map