siRNA Details
| Gene Symbol: VAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297438.1 | NP_001284367.1 | 3 | 639 | ||
| NM_014231.4 | NP_055046.1 | 3 | 639 | |||
| NM_016830.3 | NP_058439.1 | 3 | 639 | |||
| NM_199245.2 | NP_954740.1 | 3 | 639 | |||
| NR_123717.1 | 2 | 512 | ||||
| GenBank mRNA | AF060538.1 | - | 3 | 401 | ||
| AK289498.1 | - | 3 | 410 | |||
| AK296294.1 | - | 3 | 417 | |||
| AK303137.1 | - | 3 | 417 | |||
| AK309184.1 | - | 2 | 223 | |||
| BC023286.1 | - | 3 | 405 | 4943992 | ||
| BE797045.1 | - | 2 | 227 | |||
| BI838752.1 | - | 3 | 434 | |||
| BQ717992.1 | - | 3 | 398 | |||
| CR542214.1 | - | 2 | 271 | |||
| CR542231.1 | - | 2 | 271 | |||
| CR749485.1 | - | 3 | 640 | |||
| CR989458.1 | - | 3 | 393 | |||
| Z48924.1 | - | 2 | 271 | |||

VAMP1
Genomic Map