siRNA Details
| Gene Symbol: PATZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014323.2 | NP_055138.2 | 1 | 1566 | ||
| NM_032050.1 | NP_114439.1 | 1 | 1566 | |||
| NM_032051.1 | NP_114440.1 | 1 | 1566 | |||
| NM_032052.1 | NP_114441.1 | 1 | 1566 | |||
| GenBank mRNA | AB208787.1 | - | 2 | 936 | ||
| AF119256.1 | - | 1 | 1565 | |||
| AF242522.1 | - | 1 | 922 | |||
| AF254082.1 | - | 1 | 1535 | |||
| AF254083.1 | - | 1 | 1535 | |||
| AF254084.1 | - | 1 | 1535 | |||
| AF254085.1 | - | 1 | 1535 | |||
| AK291803.1 | - | 1 | 1079 | |||
| AL096880.1 | - | 1 | 900 | |||
| AY028384.1 | - | 1 | 1296 | |||
| BC021091.1 | - | 1 | 1113 | 4857537 | ||
| CR456613.1 | - | 1 | 1004 | |||

PATZ1
Genomic Map