siRNA Details
| Gene Symbol: RUSC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105203.1 | NP_001098673.1 | 10 | 3168 | ||
| NM_001105204.1 | NP_001098674.1 | 10 | 2850 | |||
| NM_001105205.1 | NP_001098675.1 | 9 | 2190 | |||
| NM_001278227.1 | NP_001265156.1 | 9 | 2134 | |||
| NM_001278228.1 | NP_001265157.1 | 8 | 2370 | |||
| NM_001278229.1 | NP_001265158.1 | 9 | 1915 | |||
| NM_001278230.1 | NP_001265159.1 | 8 | 2032 | |||
| NM_014328.4 | NP_055143.2 | 9 | 2233 | |||
| NR_103478.1 | 8 | 1580 | ||||
| XM_006711254.1 | XP_006711317.1 | 11 | 3579 | |||
| XM_006711256.1 | XP_006711319.1 | 11 | 3261 | |||
| XM_006711257.1 | XP_006711320.1 | 9 | 1847 | |||
| XM_017000891.1 | XP_016856380.1 | 10 | 3749 | |||
| XM_017000892.1 | XP_016856381.1 | 9 | 1529 | |||
| GenBank mRNA | AB026894.2 | - | 9 | 1657 | ||
| AK074982.1 | - | 9 | 1651 | |||
| AK125378.1 | - | 10 | 2802 | |||
| AK298730.1 | - | 9 | 1644 | |||
| AL080083.1 | - | 8 | 1510 | |||
| BC001045.2 | - | 9 | 1674 | |||
| BC004293.1 | - | 8 | 1436 | |||
| BX640612.1 | - | 8 | 1850 | |||
| CR749671.1 | - | 8 | 2739 | |||

RUSC1
Genomic Map