siRNA Details
| Gene Symbol: SND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014390.2 | NP_055205.2 | 3 | 513 | ||
| XM_017011987.1 | XP_016867476.1 | 3 | 542 | |||
| GenBank mRNA | AB209510.1 | - | 3 | 451 | ||
| AK096583.1 | - | 4 | 438 | |||
| AK304181.1 | - | 3 | 468 | |||
| AK312313.1 | - | 3 | 467 | |||
| BC017180.2 | - | 3 | 469 | 3345037 | ||
| BT009785.1 | - | 3 | 287 | |||
| U22055.1 | - | 3 | 479 | |||

SND1
Genomic Map