siRNA Details
| Gene Symbol: UTS2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018949.2 | NP_061822.1 | 1 | 82 | ||
| GenBank mRNA | BC136490.1 | - | 1 | 82 | 9020479 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UTS2R| Gene Symbol: UTS2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018949.2 | NP_061822.1 | 1 | 82 | ||
| GenBank mRNA | BC136490.1 | - | 1 | 82 | 9020479 | |
Set Membership: |
|